3,4-Dibromotoluene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,2-Dibromo-4-methylbenzene

D2462-25G D2462-5G
TCIAD2462-25GEA 2100 SEK
TCIAD2462-25G TCIAD2462-5G
3,4-Dibromotoluene ≥98.0%
3,4-Dibromotoluene
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79220cc6bcaba9f8d2eb51d8a821f083af8160c6&vid=073f569ad2c19671d409b5f548917a8a86d2a4dc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR