4-Allylveratrole ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol
D1360-25ML D1360-500ML
TCIAD1360-25MLEA 215 SEK
TCIAD1360-25ML TCIAD1360-500ML
4-Allylveratrole ≥98.0%
4-Allylveratrole
Formel: C₁₁H₁₄O₂
MW: 178,23 g/mol
Kokpunkt: 120°C
Smältpunkt: –4 °C
Densitet: 1,036 g/cm³ (20 °C)
Flampunkt: 130°C
Storage Temperature: Omgivande
MDL Number: MFCD00008652
CAS nummer: 93-15-2
EINECS: 202-223-0
Merck Index: 14,06073
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c733f2f3fe0bfbeffb460c04e39c2358b63c29d&vid=f99b67b4c4f49820c56c48f6a977b0c47edcf329&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %