1,3,5-Trimethoxybenzene 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
O,O,O-1,3,5-Trimethylresorcinol
P0250-25G
P0250-500G
P0250-100G
TCIAP0250-25GEA
565
SEK
TCIAP0250-25G
TCIAP0250-500G
TCIAP0250-100G
1,3,5-Trimethoxybenzene 98,0 GC_ANALYSMETOD
1,3,5-Trimethoxybenzene
Formel:
C₉H₁₂O₃ MW: 168.19 g/mol Kokpunkt: 190°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008385 CAS nummer: 621-23-8 EINECS: 210-673-4 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 53.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...