Veratrol ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether
D0627-25G D0627-500G D0627-100G
TCIAD0627-25GEA 255 SEK
TCIAD0627-25G TCIAD0627-500G TCIAD0627-100G
Veratrol ≥99.0%
Veratrol
Formel: C₈H₁₀O₂
MW: 138,17 g/mol
Kokpunkt: 207°C
Smältpunkt: 19…21 °C
Densitet: 1,085 g/cm³ (20 °C)
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008357
CAS nummer: 91-16-7
EINECS: 202-045-3
Merck Index: 14,09956
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a91b4787d9319707ae09ec9c81442a742451513&vid=0577c3913971ee04060b78057f82d18525e7e169&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %
Freezing point 21.0 to 23.0 deg-C