Ethyl isothiocyanatoacetate ≥98.0%

Leverantör: TCI
Fara

Synonyms: Ethyl 2-isothiocyanatoacetate

I0558-1G I0558-5G
TCIAI0558-1GEA 215 SEK
TCIAI0558-1G TCIAI0558-5G
Ethyl isothiocyanatoacetate ≥98.0%
Ethyl isothiocyanatoacetate
Formel: C₅H₇NO₂S
Kokpunkt: 215°C
MDL Number: MFCD00060677
CAS nummer: 24066-82-8
EINECS: 417-720-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4430dd9578167a68bfca5699343ce7b3275a5781&vid=4430dd9578167a68bfca5699343ce7b3275a5781&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR