Diglycolic anhydride ≥98.0%
Leverantör: TCI
|
Varning
|
D2308-25G
D2308-100G
TCIAD2308-25GEA
965
SEK
TCIAD2308-25G
TCIAD2308-100G
Diglycolic anhydride ≥98.0%
Diglycolic anhydride
Formel:
C₄H₄O₄ Kokpunkt: 130°C |
MDL Number:
MFCD00006677 CAS nummer: 4480-83-5 EINECS: 224-761-5 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Anilin Method) | min. 98.0 % |
Melting point | 94.0 to 98.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...