2,5-Dihydrofuran ≥98.0%

Leverantör: TCI
Fara

D0551-25ML D0551-500ML D0551-100ML
TCIAD0551-25MLEA 485 SEK
TCIAD0551-25ML TCIAD0551-500ML TCIAD0551-100ML
2,5-Dihydrofuran ≥98.0%
2,5-Dihydrofuran
Formel: C₄H₆O
Kokpunkt: 67°C
Flampunkt: 1°C
MDL Number: MFCD00003219
CAS nummer: 1708-29-8
EINECS: 216-957-4
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cf9acb08a28eb5e9bdf2fd91ab717fc618a11da&vid=2cf9acb08a28eb5e9bdf2fd91ab717fc618a11da&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR