α-Ionone ≥90.0%

Leverantör: TCI EUROPE

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

I0076-250G I0076-25G
TCIAI0076-250GEA 2900 SEK
TCIAI0076-250G TCIAI0076-25G
α-Ionone ≥90.0%
α-Ionone
Formel: C₁₃H₂₀O
MW: 192,3 g/mol
Kokpunkt: 148°C
Flampunkt: 104°C
MDL Number: MFCD00001565
CAS nummer: 127-41-3
EINECS: 204-841-6
Merck Index: 14,05056
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4d4ecf1ae7fb50e5bca83cfb1eb1a7921475e67&vid=f4d4ecf1ae7fb50e5bca83cfb1eb1a7921475e67&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR