Dimethyl bromomalonate ≥95.0%

Leverantör: TCI
Fara

D3887-25G D3887-5G
TCIAD3887-25GEA 1200 SEK
TCIAD3887-25G TCIAD3887-5G
Dimethyl bromomalonate ≥95.0%
Dimethyl bromomalonate
Formel: C₅H₇BrO₄
Kokpunkt: 225°C
MDL Number: MFCD00025865
CAS nummer: 868-26-8
EINECS: 212-775-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=effdebdc441f8464b02ae901618462033092671e&vid=effdebdc441f8464b02ae901618462033092671e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR