Dibenzyl sulphoxide ≥99.0%

Leverantör: TCI EUROPE

Synonyms: Benzyl sulphoxide, Dibenzyl sulphoxide

D0001-25G D0001-100G
TCIAD0001-25GEA 495 SEK
TCIAD0001-25G TCIAD0001-100G
Dibenzyl sulphoxide ≥99.0%
Dibenzyl sulphoxide
Formel: C₁₄H₁₄OS
MW: 230,33 g/mol
MDL Number: MFCD00004782
CAS nummer: 621-08-9
EINECS: 210-668-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=486f594a72ec68894f313850b671c39f0fd4cc3d&vid=486f594a72ec68894f313850b671c39f0fd4cc3d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 133.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR