5-Fluorooxindole ≥98.0%

Leverantör: TCI

Synonyms: 5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole

F0699-1G F0699-5G
TCIAF0699-1GEA 670 SEK
TCIAF0699-1G TCIAF0699-5G
5-Fluorooxindole ≥98.0%
5-Fluorooxindole
Formel: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD02179598
CAS nummer: 56341-41-4
EINECS: 446-540-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52a9b0deb7487c614842ccfed99d036eb431451&vid=e52a9b0deb7487c614842ccfed99d036eb431451&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR