6-Chlorooxindole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Chloro-2-oxindole

C2024-25G C2024-5G
TCIAC2024-25GEA 1225 SEK
TCIAC2024-25G TCIAC2024-5G
6-Chlorooxindole ≥98.0%
6-Chlorooxindole
Formel: C₈H₆ClNO
MW: 167,59 g/mol
MDL Number: MFCD00209962
CAS nummer: 56341-37-8
EINECS: 421-330-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fe2193478ef2d5003f9da053a8a22dbd702dd40&vid=0fe2193478ef2d5003f9da053a8a22dbd702dd40&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR