1-Benzyl-4-piperidone ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone
B1027-25ML B1027-500ML
TCIAB1027-25MLEA 295 SEK
TCIAB1027-25ML TCIAB1027-500ML
1-Benzyl-4-piperidone ≥98.0% (by GC, titration analysis)
1-Benzyl-4-piperidone
Formel: C₁₂H₁₅NO
Kokpunkt: 158°C
Flampunkt: 71°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006192
CAS nummer: 3612-20-2
EINECS: 222-782-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f1902366327f5935e6d9323fd27f851972b5e05&vid=8f1902366327f5935e6d9323fd27f851972b5e05&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %