Trimethylsilyl isocyanate ≥95.0%
Leverantör: TCI
|
Fara
|
T1106-25G
T1106-5G
T1106-100G
TCIAT1106-25GEA
1975
SEK
TCIAT1106-25G
TCIAT1106-5G
TCIAT1106-100G
Trimethylsilyl isocyanate ≥95.0%
Trimethylsilyl isocyanate
Formel:
C₄H₉NOSi Kokpunkt: 90°C Flampunkt: -2°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00001993 CAS nummer: 1118-02-1 EINECS: 214-256-8 UN: 2478 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...