4-Sulphamoylbenzoic acid ≥95.0%

Leverantör: TCI EUROPE

Synonyms: 4-Sulphamoylbenzoic acid

S0323-25G S0323-500G
TCIAS0323-25GEA 245 SEK
TCIAS0323-25G TCIAS0323-500G
4-Sulphamoylbenzoic acid ≥95.0%
4-Sulphamoylbenzoic acid
Formel: C₇H₇NO₄S
MW: 201,2 g/mol
MDL Number: MFCD00007938
CAS nummer: 138-41-0
EINECS: 205-327-4
Merck Index: 14,01876
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b57a5cef86dbd82be0b3a0371504dd54bcbacf&vid=a6b57a5cef86dbd82be0b3a0371504dd54bcbacf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR