3-Nitroanthranilic acid ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Amino-3-nitrobenzoic acid

N0879-25G N0879-5G
TCIAN0879-25GEA 5450 SEK
TCIAN0879-25G TCIAN0879-5G
3-Nitroanthranilic acid ≥97.0%
3-Nitroanthranilic acid
Formel: C₇H₆N₂O₄
MW: 182,14 g/mol
MDL Number: MFCD00024261
CAS nummer: 606-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e665e8f05166b804deebb9bd9d1ef12b8649523f&vid=e665e8f05166b804deebb9bd9d1ef12b8649523f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR