3-Methoxyphenol ≥98.0%

Leverantör: TCI
Fara

M0122-25ML M0122-500ML M0122-100ML
TCIAM0122-25MLEA 365 SEK
TCIAM0122-25ML TCIAM0122-500ML TCIAM0122-100ML
3-Methoxyphenol ≥98.0%
3-Methoxyphenol
Formel: C₇H₈O₂
Kokpunkt: 244°C
Smältpunkt: –2 °C
Densitet: 1,131 g/cm³ (20 °C)
Flampunkt: 124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002267
CAS nummer: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0ef01b8db74e92696af81c8091dac93a0c86529&vid=d95f24e3d08c214cc54f9f3f59e8a794779c0cc9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR