3,5-Bis(trifluoromethyl)thiophenol ≥96.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,5-Bis(trifluoromethyl)benzenethiol
B2391-1G
B2391-5G
TCIAB2391-1GEA
695
SEK
TCIAB2391-1G
TCIAB2391-5G
3,5-Bis(trifluoromethyl)thiophenol ≥96.0% (by GC, titration analysis)
3,5-Bis(trifluoromethyl)thiophenol
Formel:
C₈H₄F₆S MW: 246,18 g/mol Flampunkt: 65°C |
MDL Number:
MFCD00042273 CAS nummer: 130783-02-7 ADR: 6.1,III |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Iodometric Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...