Diethyl diethylmalonate ≥97.0%

Leverantör: TCI

D0867-25ML D0867-500ML
TCIAD0867-25MLEA 375 SEK
TCIAD0867-25ML TCIAD0867-500ML
Diethyl diethylmalonate ≥97.0%
Diethyl diethylmalonate
Formel: C₁₁H₂₀O₄
Flampunkt: 94°C
MDL Number: MFCD00009132
CAS nummer: 77-25-8
EINECS: 201-016-2
Merck Index: 14,03823
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098b350ac12a0f7f1e008fb03c0ed58cd14467cc&vid=098b350ac12a0f7f1e008fb03c0ed58cd14467cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR