1,2,3,4-Tetrahydroquinoline ≥95.0%

Leverantör: TCI EUROPE
Varning

T0113-25ML T0113-500ML T0113-100ML
TCIAT0113-25MLEA 215 SEK
TCIAT0113-25ML TCIAT0113-500ML TCIAT0113-100ML
1,2,3,4-Tetrahydroquinoline ≥95.0%
1,2,3,4-Tetrahydroquinoline
Formel: C₉H₁₁N
MW: 133,19 g/mol
Kokpunkt: 251°C
Flampunkt: 113°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006693
CAS nummer: 635-46-1
EINECS: 211-237-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=832cdada89b3cf8cff167e218bff97534a2868ad&vid=832cdada89b3cf8cff167e218bff97534a2868ad&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 95.0 %
Freezing point 11.0 to 20.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR