Disodium malonate ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
Sodium malonate dibasic, Malonic acid disodium salt
M0031-25G
M0031-500G
TCIAM0031-25GEA
455
SEK
TCIAM0031-25G
TCIAM0031-500G
Disodium malonate ≥99.0%
Disodium malonate
Formel:
C₃H₂Na₂O₄ MW: 148,03 g/mol |
MDL Number:
MFCD00002708 CAS nummer: 141-95-7 EINECS: 205-514-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...