4-Ethoxyphenol ≥98.0%

Leverantör: TCI EUROPE
Varning

E0216-25G E0216-500G
TCIAE0216-25GEA 725 SEK
TCIAE0216-25G TCIAE0216-500G
4-Ethoxyphenol ≥98.0%
4-Ethoxyphenol
Formel: C₈H₁₀O₂
MW: 138,17 g/mol
Kokpunkt: 121°C
Smältpunkt: 63…64 °C
MDL Number: MFCD00002334
CAS nummer: 622-62-8
EINECS: 210-748-1

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR