3-Ethoxyphenol ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
E0051-25ML
E0051-5ML
TCIAE0051-25MLEA
3025
SEK
TCIAE0051-25ML
TCIAE0051-5ML
3-Ethoxyphenol ≥97.0%
3-Ethoxyphenol
Formel:
C₈H₁₀O₂ MW: 138,17 g/mol Kokpunkt: 247°C Densitet: 1,105 g/cm³ (15 °C) |
MDL Number:
MFCD00016450 CAS nummer: 621-34-1 EINECS: 210-681-8 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow to Light orange powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...