Triphenylamine ≥98.0%

Leverantör: TCI
Varning

T0507-25G T0507-250G
TCIAT0507-25GEA 430 SEK
TCIAT0507-25G TCIAT0507-250G
Triphenylamine ≥98.0%
Triphenylamine
Formel: C₁₈H₁₅N
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 123…128 °C
Densitet: 0,774 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003020
CAS nummer: 603-34-9
EINECS: 210-035-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=598aee6414624881a8a0433b2445fdc074f6cbff&vid=8f7e54d3e61abd8d775dd33506fff8539ee88910&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR