N-Metylanilin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
M0147-25G M0147-500G
TCIAM0147-25GEA 255 SEK
TCIAM0147-25G TCIAM0147-500G
N-Metylanilin ≥98.0% (by GC, titration analysis)
N-Metylanilin
Formel: C₇H₉N
MW: 107.16 g/mol
Kokpunkt: 196°C
Smältpunkt: –57°C
Flampunkt: 83°C
Storage Temperature: Omgivande
MDL Number: MFCD00008283
CAS nummer: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1 III
Merck Index: 14,06019
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=978691bad2b138bcad1fbb38c1a9261c8c53241f&vid=7546da49c5448b0f014c8b1f6a76ae3ced97f402&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %