N-Etylanilin ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: N-Ethyl-N-phenylamine
E0059-25ML E0059-500ML
TCIAE0059-25MLEA 215 SEK
TCIAE0059-25ML TCIAE0059-500ML
N-Etylanilin ≥99.0% (by GC, titration analysis)
N-Etylanilin
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 204°C
Smältpunkt: –64 °C
Densitet: 0,96 g/cm³ (20 °C)
Flampunkt: 91°C
Storage Temperature: Omgivande
MDL Number: MFCD00009025
CAS nummer: 103-69-5
EINECS: 203-135-5
UN: 2272
ADR: 6.1,III
Merck Index: 14,03764
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192d15c08a8d9db012d47e9d706341363223a57a&vid=149875befb9b84920c90e0fa8edfc72777505080&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %