2,5-Dimethoxyaniline ≥98.0%
Leverantör: TCI EUROPE
Fara
D1893-25G D1893-500G
TCIAD1893-25GEA 295 SEK
TCIAD1893-25G TCIAD1893-500G
2,5-Dimethoxyaniline ≥98.0%
2,5-Dimethoxyaniline
Formel: C₈H₁₁NO₂
MW: 153,18 g/mol
Kokpunkt: 159°C
Smältpunkt: 81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008368
CAS nummer: 102-56-7
EINECS: 203-040-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d76b81a354daa3c0249fc0c011c3e5b8cd349b0&vid=dc77e8e93e4890c586c199b3bedd7dce3610d70f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 78.0 to 82.0 deg-C