3,5-Dinitroanilin ≥98.0%
Leverantör: TCI EUROPE
Fara
D2458-25G D2458-5G
TCIAD2458-25GEA 3525 SEK
TCIAD2458-25G TCIAD2458-5G
3,5-Dinitroanilin ≥98.0%
3,5-Dinitroanilin
Formel: C₆H₅N₃O₄
MW: 183,12 g/mol
Smältpunkt: 163 °C
Densitet: 1,601 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007263
CAS nummer: 618-87-1
EINECS: 210-567-8
UN: 1596
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38df2df028708e1243ffe535ec6680f956104949&vid=7c3d8eb9929f90f93923942b2c35694518692b30&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in Methanol almost transparency