4-Nitroanilin ≥98.0%
Leverantör: TCI
Fara
Synonyms: p-Nitroanilin
N0119-25G N0119-500G
TCIAN0119-25GEA 215 SEK
TCIAN0119-25G TCIAN0119-500G
4-Nitroanilin ≥98.0%
4-Nitroanilin
Formel: C₆H₆N₂O₂
Kokpunkt: 332°C
Smältpunkt: 148 °C
Densitet: 1,24 g/cm³ (20 °C)
Flampunkt: 199 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007858
CAS nummer: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1,II
Merck Index: 14,06584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=004b7dd75e92352cf4b7e738e089a060d9ceea37&vid=477c79e71f2ffbb78f5cf0ac8ccb29d032461a18&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 150.0 deg-C
Solubility in hot EtOH almost transparency