3-Nitroanilin ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: m-Nitroanilin

N0117-25G N0117-500G
TCIAN0117-25GEA 445 SEK
TCIAN0117-25G TCIAN0117-500G
3-Nitroanilin ≥98.0%
3-Nitroanilin
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Kokpunkt: 306 °C (1013 hPa)
Smältpunkt: 109…112 °C
Densitet: 0,9 g/cm³ (25 °C)
Flampunkt: 196 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007782
CAS nummer: 99-09-2
EINECS: 202-729-1
UN: 1661
ADR: 6.1,II
Merck Index: 14,06581
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ceb58a09f61a69eda611bdd5e199ca45e532b71&vid=25f34aca72e9a59a7b60d364f43d35ef87b73b02&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 114.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR