2-Nitroanilin ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: o-Nitroanilin

N0118-25G N0118-500G
TCIAN0118-25GEA 215 SEK
TCIAN0118-25G TCIAN0118-500G
2-Nitroanilin ≥99.0%
2-Nitroanilin
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Kokpunkt: 165°C
Smältpunkt: 68…71 °C
Densitet: 1,446 g/cm³ (20 °C)
Flampunkt: 167 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007687
CAS nummer: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1,II
Merck Index: 14,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0c27a4faf4186b3263503da7e420245721400b8&vid=1734a30a65418988b3791f95784a14ef0f2d4d20&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 72.0 to 74.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR