N,N-Dibutylaniline ≥98.0%

Leverantör: TCI EUROPE
Varning

D0222-25ML D0222-500ML D0222-100ML
TCIAD0222-25MLEA 605 SEK
TCIAD0222-25ML TCIAD0222-500ML TCIAD0222-100ML
N,N-Dibutylaniline ≥98.0%
N,N-Dibutylaniline
Formel: (C₆H₅)N(CH₂CH₂CH₂CH₃)₂
MW: 205,34 g/mol
Kokpunkt: 266°C
Smältpunkt: –32 °C
Densitet: 0,90368 g/cm³ (20 °C)
MDL Number: MFCD00035799
CAS nummer: 613-29-6
EINECS: 210-335-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=591854da6ffbbef8635dced8a9b361486cc58b88&vid=6f71ade11a0cae7f4e3d38dc604afd132d8d9f06&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR