3-Iodophenol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
I0495-10G
I0495-25G
TCIAI0495-10GEA
885
SEK
TCIAI0495-10G
TCIAI0495-25G
3-Iodophenol ≥98.0%
3-Iodophenol
Formel:
C₆H₅IO MW: 220,01 g/mol Kokpunkt: 190°C |
MDL Number:
MFCD00002261 CAS nummer: 626-02-8 EINECS: 210-923-2 ADR: 9, |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...