2-Iodophenol ≥99.0%
Leverantör: TCI EUROPE
Varning
I0460-25G I0460-5G
TCIAI0460-25GEA 1475 SEK
TCIAI0460-25G TCIAI0460-5G
2-Iodophenol ≥99.0%
2-Iodophenol
Formel: C₆H₅IO
MW: 220,01 g/mol
Kokpunkt: 187°C
Smältpunkt: 39…42 °C
Densitet: 1,8757 g/cm³ (80 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00013963
CAS nummer: 533-58-4
EINECS: 208-569-9
ADR: 6.1,III
Merck Index: 14,05035
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=817ea8833c0e8fbccc53403bfa627b3740adf7c6&vid=6e75eb771177018a50b66c05693038cced40988b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 99.0 %
Melting point 40.0 to 44.0 deg-C
Solubility in Methanol almost transparency