4-Iodoanisole ≥99.0%
Leverantör: TCI EUROPE
Varning
I0378-25G I0378-100G
TCIAI0378-25GEA 325 SEK
TCIAI0378-25G TCIAI0378-100G
4-Iodoanisole ≥99.0%
4-Iodoanisole
Formel: C₇H₇IO
MW: 234,04 g/mol
Kokpunkt: 240°C
Smältpunkt: 48…49 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001056
CAS nummer: 696-62-8
EINECS: 211-798-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2534f409753b95ee624f56a060ead3f910ae66&vid=06873d3ee9e7e8ec7dbed6b5b1bc00b320343972&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 49.0 to 52.0 deg-C
Solubility in Methanol almost transparency