2-Iodoaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
I0046-25G I0046-5G
TCIAI0046-25GEA 910 SEK
TCIAI0046-25G TCIAI0046-5G
2-Iodoaniline ≥98.0% (by GC, titration analysis)
2-Iodoaniline
Formel: C₆H₆IN
MW: 219,02 g/mol
Kokpunkt: 263 °C (1013 hPa)
Smältpunkt: 54…57 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007680
CAS nummer: 615-43-0
EINECS: 210-426-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff530ada4776ad57a4996edb867b2d649644bac5&vid=2b0f660c3448471ee9529af77dd6de9aab78ccfc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 56.0 to 59.0 deg-C
Solubility in Methanol very faint turbidity