4-Iodobenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: p-Iodobenzoic acid
I0054-25G I0054-5G
TCIAI0054-25GEA 725 SEK
TCIAI0054-25G TCIAI0054-5G
4-Iodobenzoic acid ≥98.0% (by GC, titration analysis)
4-Iodobenzoic acid
Formel: C₇H₅IO₂
MW: 248.02 g/mol
Kokpunkt: 319
Storage Temperature: Omgivande
MDL Number: MFCD00002533
CAS nummer: 619-58-9
EINECS: 210-603-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d4df5d48e8676d65c091cd27205b7792b13671&vid=c8d4df5d48e8676d65c091cd27205b7792b13671&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity