3-Iodobenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: m-Iodobenzoic acid
I0052-25G I0052-5G
TCIAI0052-25GEA 895 SEK
TCIAI0052-25G TCIAI0052-5G
3-Iodobenzoic acid ≥98.0% (by GC, titration analysis)
3-Iodobenzoic acid
Formel: C₇H₅IO₂
MW: 248,02 g/mol
Kokpunkt: 337 °C (1013 hPa)
Smältpunkt: 186…188 °C
Densitet: 2,171 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002496
CAS nummer: 618-51-9
EINECS: 210-555-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd22f00c02f374cde694dff67ca49a01d7f3f466&vid=b03fe9225d4e27b17e6fef0273d498c9b56a995a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Solubility in Methanol almost transparency