1,4-Diiodobenzene ≥98.0%
Leverantör: TCI EUROPE
D0608-25G
D0608-5G
TCIAD0608-25GEA
870
SEK
TCIAD0608-25G
TCIAD0608-5G
1,4-Diiodobenzene ≥98.0%
1,4-Diiodobenzene
Formel:
C₆H₄I₂ MW: 329,91 g/mol Kokpunkt: 285 °C (1013 hPa) Smältpunkt: 126…127 °C Densitet: 2,88 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00001054 CAS nummer: 624-38-4 EINECS: 210-842-2 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 and le 132.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...