2,3,5,6-Tetrafluorophenol ≥98.0%

Leverantör: TCI EUROPE
Varning

T1030-25G T1030-5G
TCIAT1030-25GEA 2275 SEK
TCIAT1030-25G TCIAT1030-5G
2,3,5,6-Tetrafluorophenol ≥98.0%
2,3,5,6-Tetrafluorophenol
Formel: C₆H₂F₄O
MW: 166,07 g/mol
Kokpunkt: 146°C
Smältpunkt: 30 °C
MDL Number: MFCD00002157
CAS nummer: 769-39-1
EINECS: 212-209-6
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a80cf0deb9a4d62231d780cad632bc6c4b544424&vid=d55fb615668aa10686012ab03afaaca8b0db5499&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR