2,3,4-Trifluorophenol ≥98.0%
Leverantör: TCI EUROPE
Fara
T1616-1G T1616-5G
TCIAT1616-1GEA 590 SEK
TCIAT1616-1G TCIAT1616-5G
2,3,4-Trifluorophenol ≥98.0%
2,3,4-Trifluorophenol
Formel: C₆H₃F₃O
MW: 148,08 g/mol
Kokpunkt: 69 °C (43 -rr)
Smältpunkt: 30…34 °C
Densitet: 1,46 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00042426
CAS nummer: 2822-41-5
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ec6d8e1a1da945ec43500f701baae6233119be8&vid=73cdd71c1bce88fd99ef06c306227541dcd1035b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %