3-Fluorophenol ≥98.0%
Leverantör: TCI
Varning
F0159-100ML F0159-25ML
TCIAF0159-100MLEA 2450 SEK
TCIAF0159-100ML TCIAF0159-25ML
3-Fluorophenol ≥98.0%
3-Fluorophenol
Formel: C₆H₅FO
Kokpunkt: 178°C
Densitet: 1,24 g/cm³ (20 °C)
Flampunkt: 71°C
Storage Temperature: Omgivande
MDL Number: MFCD00002254
CAS nummer: 372-20-3
EINECS: 206-748-6
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23352bc7a0078cdbeeece4255520b0400a147e4e&vid=d7b17be26e51f99094d72be48e03593843101a67&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %