2-Fluoro-6-nitrophenol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Fluoro-2-nitrophenol

F0623-1G F0623-5G
TCIAF0623-1GEA 575 SEK
TCIAF0623-1G TCIAF0623-5G
2-Fluoro-6-nitrophenol ≥98.0%
2-Fluoro-6-nitrophenol
Formel: C₆H₄FNO₃
MW: 157,1 g/mol
Smältpunkt: 91…92 °C
MDL Number: MFCD00042446
CAS nummer: 1526-17-6
EINECS: 216-199-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b483813393e5c9aaccb7874eddac4fa3484953a&vid=f6a83ce56c5d14d16172126b7fc3970d122f6b29&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR