3,5-Difluorophenol ≥98.0%

Leverantör: TCI EUROPE
Varning

D2569-25G D2569-5G
TCIAD2569-25GEA 2425 SEK
TCIAD2569-25G TCIAD2569-5G
3,5-Difluorophenol ≥98.0%
3,5-Difluorophenol
Formel: C₆H₄F₂O
MW: 130,09 g/mol
Kokpunkt: 68°C
Smältpunkt: 55 °C
Densitet: 1,566 g/cm³ (20 °C)
MDL Number: MFCD00002255
CAS nummer: 2713-34-0
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ecc12601826ee0239a2fe5b4fa6c395219c428&vid=8fc083419e1e5ee1e2cdc958bcc9d24191e4f35a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 53.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR