3,4-Difluoroanisole ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
D2856-25G
D2856-5G
TCIAD2856-25GEA
1825
SEK
TCIAD2856-25G
TCIAD2856-5G
3,4-Difluoroanisole ≥97.0%
3,4-Difluoroanisole
Formel:
C₇H₆F₂O MW: 144,12 g/mol Kokpunkt: 139 °C (1013 hPa) Smältpunkt: –30 °C Densitet: 1,23 g/cm³ (20 °C) Flampunkt: 55°C |
MDL Number:
MFCD00010730 CAS nummer: 115144-40-6 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...