4-Fluoro-3-nitroaniline ≥98.0%

Leverantör: TCI EUROPE
Varning

F0168-10G F0168-25G
TCIAF0168-10GEA 295 SEK
TCIAF0168-10G TCIAF0168-25G
4-Fluoro-3-nitroaniline ≥98.0%
4-Fluoro-3-nitroaniline
Formel: C₆H₅FN₂O₂
MW: 156,12 g/mol
Smältpunkt: 96…98 °C
Densitet: 0,43 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007833
CAS nummer: 364-76-1
EINECS: 206-665-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bec1d4aadd2f52a5caa7f99c658f930ab2aaa59&vid=e692d350300f0ad292d6fab1c9cc2563b47a41bc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 98.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR