4-Fluoro-2-nitroaniline ≥98.0%
Leverantör: TCI EUROPE
Varning
F0380-25G F0380-5G
TCIAF0380-25GEA 2075 SEK
TCIAF0380-25G TCIAF0380-5G
4-Fluoro-2-nitroaniline ≥98.0%
4-Fluoro-2-nitroaniline
Formel: C₆H₅FN₂O₂
MW: 156,12 g/mol
Smältpunkt: 95 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007830
CAS nummer: 364-78-3
EINECS: 206-666-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e427ae27e6b33d5c9ad7efecf5ec18b683799c6&vid=cd96268901b3894395ea5b10fe4fb85b516c9675&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in Methanol very faint turbidity