2,3,5,6-Tetrafluoroaniline ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
T1028-25G
T1028-5G
TCIAT1028-25GEA
2375
SEK
TCIAT1028-25G
TCIAT1028-5G
2,3,5,6-Tetrafluoroaniline ≥97.0%
2,3,5,6-Tetrafluoroaniline
Formel:
C₆H₃F₄N MW: 165,09 g/mol Kokpunkt: 158°C Smältpunkt: 31…32 °C Densitet: 1,744 g/cm³ (20 °C) Flampunkt: 62°C |
MDL Number:
MFCD00007644 CAS nummer: 700-17-4 EINECS: 211-840-4 ADR: 6.1,III |
Specifikation testresultat
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...