2,3,4-Trifluoroanilin ≥98.0%

Leverantör: TCI EUROPE
Fara

T1385-25G T1385-5G
TCIAT1385-25GEA 1150 SEK
TCIAT1385-25G TCIAT1385-5G
2,3,4-Trifluoroanilin ≥98.0%
2,3,4-Trifluoroanilin
Formel: C₆H₄F₃N
MW: 147,1 g/mol
Kokpunkt: 92°C
Smältpunkt: 14…15 °C
Densitet: 1,39 g/cm³ (20 °C)
Flampunkt: 68°C
MDL Number: MFCD00011737
CAS nummer: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0b840f556f602f97bfe76a045305252f2941a4&vid=d90d402378191ad7b616364cf73b686abf6b5270&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR