4-Fluoro-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

F0399-25G F0399-5G
TCIAF0399-25GEA 1075 SEK
TCIAF0399-25G TCIAF0399-5G
4-Fluoro-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-3-nitrobenzoic acid
Formel: C₇H₄FNO₄
Kokpunkt: 92 °C (15 -rr)
Smältpunkt: 123…124 °C
MDL Number: MFCD00007058
CAS nummer: 453-71-4
EINECS: 207-221-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=570cca0830bf66b0e1681a6c64b6a17dd7c04b30&vid=633cf7758b001ab9a0cb5842b0b14a6b9dc3fdd1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR